Would you like to log in again?
HABITAT Lid Stands for HABITAT research Bioreactors
Supplier: IKA
35004067 35004059
76836-718EA 1848 USD InStock
76836-718 76836-682
HABITAT Lid Stands for HABITAT research Bioreactors
Bioreactors
This sturdy lid stand can be used for 2, 5 and 10 L glass vessels.
You can conveniently hang the lid on the stand, so that the individual components can be easily mounted or dismounted. In addition, you have the option to put the motor and sensors in designated places, so that the workplace remains tidy. The pressure spring tripod rod also makes it easy to move the tripod up and down.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty