Would you like to log in again?
Liquid Chromatograph with UV Detector
Supplier: Lucidity Systems
928274
76836-906EA 36225 USD InStock
76836-906
Liquid Chromatograph with UV Detector
Chromatography Systems Complete HPLC Systems
High Performance Liquid Chromatograph with UV detector.
  • LC-UV with integrated multi-wavelength UV detector
  • Equipped with a 20 place autosampler
  • Binary pump system
  • High pressure HPLC pumps for gradient flow up to 10000 psi
  • Integrated degassers and temperature programmable column oven
  • Includes laptop with Lucidity Software
The Lucidity LC-UV is one of the smallest, most economical, and robust HPLCs on the market. Equipped with a variable wavelength UV detector capable of measuring signal from 185 nm to 475 nm, the Lucidity LC-UV has a binary gradient pumping system existing of two high pressure piston pumps that deliver accurate solvent gradients up to 10000 psi.
With a 20-position autosampler and integrated degassers the Lucidity LC-UV has everything required to provide great chromatography results.
Delivery information: The instrument comes with everything required for analysis, including: laptop containg Lucidity Software, starter column, full accessory kit and training videos. Self-installable in under 30 minutes. With a 24-month warranty, the LC-UV will provide reliable results for years to come.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty