Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
UVP BioSpectrum Advanced Bioimaging Systems
Supplier: Analytik Jena
849-97-0945-01 849-97-0945-03 849-97-0946-01 849-97-0945-02 849-97-0945-04 849-97-0946-02
76837-116EA 145210.08 USD InStock
76837-116 76837-114 76837-094 76837-108 76837-120 76837-096
UVP BioSpectrum Advanced Bioimaging Systems
Photodocumentation Systems
The all-in-one imager for any imaging application. From gel analysis and documentation to in vivo fluorescence and bioluminescence animal imaging, the UVP Biospectrum Advanced does it all.
  • Two high performance cooled camera options
  • Illumination sources - overhead laser bars in RGBW, NIR, and violet (405 nm)
  • Motorized lift for sample magnification range
  • Included warming plate or UV transilluminator
  • VisionWorks software for one-click workflows
The UVP BioSpectrum Advanced is an all in-one BioImaging system with in vivo imaging capabilities for sample documentation and analyzes, including in vivo animal imaging, plant imaging, chemiluminescence, NIR, and fluorescence. This all-in-one system is perfect for any lab within research institutions, medical facilities, academic research institutions, pharmaceutical companies and more.
The UVP BioSpectrum Advanced comes with a choice of deeply-cooled CCD camera sensors capable of imaging fluorescence, chemiluminescence, and bioluminescence. The system features extended NIR, violet (405 nm), red, green, and blue laser bars, white epi (overhead) illumination, motorized platform lift and filter wheel, and automated lens. Add a sample warming plate, UV light transilluminator, or blue light transilluminator to fit your specific application needs.
Delivery information: Included with system purchase depending on in vivo or fluorescence/chemiluminescence configurations: GFP and RFP emission filters (included with in vivo configuration) or EtBr emission filter (included with fluorescence/chemiluminescence configuration). Warming plate for 1 to 10 mice (included with in vivo configuration) or UVP Transilluminator, 302 nm (included with fluorescence/chemiluminescence configuration). VisionWorks Software for image capture, enhancement and analyzes (unlimited licenses for both configurations).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty