Would you like to log in again?
Glass Jugs, Amber
Supplier: Qorpak
323664 259531 GLC-02043 307412 GLC-02057 GLC-13300 GLA-00907 GLA-08344 259534 GLC-02054 GLC-07021 GLC-07065 259354 GLA-00906 GLC-02055 323201 GLC-02065 325552 GLC-02050 GLC-04890 GLC-02045 GLC-02046
10770-404CS 94.17 USD InStock
10770-404 10770-318 76487-208 93000-934 16142-468 16142-402 76848-010 89204-947 76500-060 16194-105 76474-074 77200-800 36319-743 76848-014 36319-786 16142-583 76487-210 18087-099 16087-018 76487-348 16087-025 77530-946
Glass Jugs, Amber
Bottles Carboys
Amber glass jugs with molded finger grips are the perfect storage solution for light sensitive products.
Amber glass protects against harmful UV rays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty