Would you like to log in again?
Rental Sampling Kits
Supplier: Stat Peel
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
A00556 A00555 A00557
76848-038EA 6528 USD InStock
76848-038 76848-044 76848-034
Rental Sampling Kits
Samplers Air Samplers
Sampling kits allow for the collection of respirable, inhalable, or both respirable and inhalable particles for inspection in the STAT PEEL® Identifier System.
  • Battery powered badges for particle collection
  • Tablet computer for badge control
  • Badges chargers
  • All necessary tools to perform air samplings
  • Starter kit of 20 filtration slides
The sampling kit allows for the collection of respirable, inhalable, or both respirable and inhalable particles for inspection in the STAT PEEL® Identifier system or for SEM inspection. The included filtration badges are personal sampling devices equipped with a silent pump, environmental sensors, and filtration slides that collect respirable particles. It can be worn in a lab coat pocket to sample personal exposure or set up in a desired location to sample environmental contamination. The badge is operated with an easy-to-use app on a tablet computer.
Each kit contains all the necessary items and instructions for the user to perform an aerosol sampling. After completion of the sampling, the kit is returned to Stat Peel for filter analysis and an exposure report.
Certifications: CE marked.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty