Would you like to log in again?
Flow Cell Priming Kit
Supplier: Oxford Nanopore Technologies
EXP-FLP004-XL EXP-FLP002 EXP-FLP004
76848-056EA 180 USD InStock
76848-056 76487-130 76636-302
Flow Cell Priming Kit
Nucleic Acid Reagents Next Generation Sequencing Reagents
Additional flow cell priming reagents to enable additional loading of more library during a sequencing run.
  • The kit can be used to prime a MinION™ Flow Cell for sequencing or to add more fuel to a flow cell during a sequencing experiment.
Flow Cell Priming Kit V14 (EXP-FLP004) ― the Flow Cell Priming Kit V14 contains Flow Cell Flush (FCF) and Flow Cell Tether (FCT), sufficient for 6 reactions. This kit is compatible with the latest library preparation kit chemistry (V14) and R10.4.1 Flow Cells.
Note: This kit is not compatible with the Ultra-Long DNA Sequencing Kit V14, which requires specific flow cell priming reagents.
Flow Cell Priming Kit (EXP-FLP002) ― the Flow Cell Priming Kit contains Flush Buffer (FB) and Flush Tether (FLT), sufficient for six reactions using R9.4.1 Flow Cells. This kit is anticipated to be discontinued in September 2023. It is recommended to transition to the latest library preparation kit chemistry (V14) and R10.4.1 Flow Cells, which are compatible with the Flow Cell Priming Kit V14.
Delivery information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Order Now
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty