Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FTO Rabbit Polyclonal Antibody

Supplier: Antibodies.com LLC
A8927-100 A8927-50
76863-682EA 225 USD InStock
76863-682 76862-110
Anti-FTO Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to FTO for WB and IP with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IP
  • Recommended Dilutions: WB: 1:500 to 1:1000, IP: 1:500 to 1:100

Type: Primary
Antigen: FTO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty