Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Avantor 3D Mixing Bags

Supplier: Avantor Fluid Handling
1010336 1010331 1010334 1010333 1010330 1010337 1010332 1010335
76894-054EA 1432.53 USD InStock
76894-054 76894-044 76894-052 76894-042 76894-050 76894-060 76894-058 76894-048
Avantor 3D Mixing Bags
Bags Media Bags
The Magnetic Mixing bags offer a sterile, single use mixing container designed to maximize mixing performance while minimizing disruptive shear.

  • 3D Avantor mixing bags with 3×50 cm 1×5 cm silicone platinum ports
  • Top Port 1: ⅜’’ port, with pinch clamp, and 1 male MPX
  • Top Port 2: ⅜’’ port, with pinch clamp, and 1 female MPX
  • Top Port 3: ¼” port, plugged.
  • Bottom Port: ⅜’’ port, with pinch clamp, and 1 female MPC
  • Top Powder Port: 4", with bioclamp and TC end cap

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty