Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium chloride solution 1 M, in single-use containers, HyClone™ for bioprocessing

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: HaliteSea saltSaline

SH31140.01 SH31140.06 SH31140.06 SH31140.02 SH31140.04 SH31140.04 SH31140.05 SH31140.02 SH31140.03 SH31140.03 SH31140.05 SH31140.01
76894-154EA 1385 USD InStock
76894-154 CA76894-170 76894-170 CA76894-118 CA76894-148 76894-118 CA76894-102 76894-148 CA76894-136 76894-102 76894-136 CA76894-154
Sodium chloride solution 1 M, in single-use containers, HyClone™ for bioprocessing
Sodium chloride
HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 µm sterile filtered.
  • Provided in single-use bag containers.
  • Raw material is EP, USP, and JP grade.

These products are manufactured upon order (100 L minimum order quantity). HyClone™ NaCl in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 1 M and 2 M sodium chloride concentrations up to 200 L. Contact us for information around bag configurations or custom bulk formulations.


Formula: NaCl
MW: 58.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Order Now
Specification Test Results
Storage Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
Endotoxin Concentration ≤ 1 EU/mL
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 months
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty