Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide solution 1 N (1 M), in single-use containers, HyClone™ for bioprocessing

Supplier: Cytiva
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

SH31088.02 SH31088.06 SH31088.05 SH31088.04 SH31088.05 SH31088.01 SH31088.03 SH31088.06 SH31088.04 SH31088.01 SH31088.02 SH31088.03
76894-110EA 818 USD InStock
76894-110 CA76894-128 76894-128 CA76894-138 CA76894-114 76894-138 CA76894-156 76894-114 CA76894-112 CA76894-110 76894-112 76894-156
Sodium hydroxide solution 1 N (1 M), in single-use containers, HyClone™ for bioprocessing
Sodium hydroxide
HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean your chromatography resins.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.1 µm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.

HyClone™ NaOH in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 0.1 M (0.1 N), 0.5 M (0.5 N), and 1.0 M (1.0 N) sodium hydroxide concentrations up to 200 L.


Formula: NaOH
MW: 40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
REACH: 01-2119457892-27
Order Now
Specification Test Results
Storage 15° to 30°C
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 months
pH For Information Only
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty