Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
SH31088.02
SH31088.06
SH31088.05
SH31088.04
SH31088.05
SH31088.01
SH31088.03
SH31088.06
SH31088.04
SH31088.01
SH31088.02
SH31088.03
76894-110EA
818
USD
InStock
76894-110
CA76894-128
76894-128
CA76894-138
CA76894-114
76894-138
CA76894-156
76894-114
CA76894-112
CA76894-110
76894-112
76894-156
Sodium hydroxide solution 1 N (1 M), in single-use containers, HyClone™ for bioprocessing
Sodium hydroxide
HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean your chromatography resins.
- Manufactured in ISO 9001 and ISO 13485 certified facility.
- Lot-specific certificate of analysis (CoA) for each lot produced.
- Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
- 0.1 µm sterile filtered.
- All unit sizes are filled in single-use, ready-to-use bags.
- Raw materials are USP/NF and EP compendial grade.
HyClone™ NaOH in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 0.1 M (0.1 N), 0.5 M (0.5 N), and 1.0 M (1.0 N) sodium hydroxide concentrations up to 200 L.