Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanol solution 20%, in single-use containers, HyClone™ for bioprocessing

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Warning
Synonyms: Ethyl alcohol

SH30926.07 SH30926.01 SH30926.06 SH30926.07 SH30926.02 SH30926.02 SH30926.05 SH30926.01 SH30926.03 SH30926.03 SH30926.05 SH30926.04 SH30926.04 SH30926.06
76894-108EA 737 USD InStock
CA76894-108 76894-108 76894-098 76894-142 CA76894-130 76894-130 CA76894-104 CA76894-116 76894-104 CA76894-146 76894-116 76894-146 CA76894-142 CA76894-098
Ethanol solution 20%, in single-use containers, HyClone™ for bioprocessing
Ethanol
HyClone 20% ethanol solution in single-use bags offers a convenient format for cleaning and storing chromatography resins in downstream bioprocessing applications.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.2 µm sterile filtered.
  • Provided in single-use bag containers.

These products are manufactured upon order (100 L minimum order quantity). HyClone 20% ethanol in single-use bags is well-suited for use with Cytiva's chromatography resins.
20% ethanol bulk liquid is available in volumes up to 200 L. Contact us for information around bag configurations or custom bulk formulations.


Formula: C₂H₅OH
MW: 46.07 g/mol
Flash Pt: 30 - 38°C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
Order Now
Specification Test Results
Storage Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU)
Endotoxin Concentration ≤ 1 EU/mL
Certifications/Compliance Manufactured in a ISO9001 and ISO13485 certified facility
Shelf life 12 months
For use with Chromatography resins, IC systems, ÄKTA hardware systems
Appearance Clear, colorless solution
Sterility No Growth

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty