Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water, HyClone™ HyPure USP, WFI (water for injection) quality

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Synonyms: Dihydrogen oxide

SH31191.02 SH31191.03 SH31191.LS SH31191.02 SH31191.01 SH31191.04 SH31191.03 SH31191.01 SH31191.LS SH31191.04
76538-682EA 28.26 USD InStock
76538-682 CA76538-680 76538-680 CA76894-126 CA76538-678 CA76538-676 76538-678 76894-126 76538-676 CA76538-682
Water, HyClone™ HyPure USP, WFI (water for injection) quality
Water
Support cell culture growth and biomanufacturing processes with Water for Injection (WFI) quality water.

  • Meets stringent USP specifications
  • Manufactured in ISO 9001 certified facility
  • 0.1 µm sterile filtered
  • Shelf life 24 months

HyClone HyPure WFI Quality Water undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP). Water for injection (WFI) is produced in ISO 9001 certified facilities to ensure a high quality product.

HyPure WFI Quality Water meets the following criteria: Must be produced from a validated water purification system that meets or exceeds current USP requirements for HyPure WFI Quality Water. Must have a quality assurance monitoring system that ensures continuous compliance and incorporates system test and action limits. Must meet all applicable chemical, purity, and other acceptance criteria as outlined in the current USP for both system and packaged water. Must include a lot-specific CoA for each lot of water produced, listing both acceptance criteria and actual test results.

Caution: Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.


Formula: H₂O
MW: 18.02 g/mol
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Order Now

 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty