Synonyms:
Halite
, Sea salt
, Saline
SH31141.03
SH31141.02
SH31141.06
SH31141.06
SH31141.05
SH31141.01
SH31141.04
SH31141.04
SH31141.01
SH31141.02
SH31141.05
SH31141.03
76894-120EA
540
USD
InStock
76894-120
76894-132
CA76894-172
76894-172
CA76894-168
CA76894-122
76894-168
CA76894-100
CA76894-120
76894-100
76894-122
CA76894-132
Sodium chloride solution 2 M, in single-use containers, HyClone™ for bioprocessing
Sodium chloride
HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.
- Manufactured in ISO 9001 and ISO 13485 certified facility.
- Lot-specific certificate of analysis (CoA) for each lot produced.
- Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
- 0.2 µm sterile filtered.
- Provided in single-use bag containers.
- Raw material is EP, USP, and JP grade.
These products are manufactured upon order (100 L minimum order quantity). HyClone™ NaCl in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 1 M and 2 M sodium chloride concentrations up to 200 L. Contact us for information around bag configurations or custom bulk formulations.