Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
SH30883.06
SH30883.07
SH30883.03
SH30883.05
SH30883.01
SH30883.06
SH30883.03
SH30883.04
SH30883.05
SH30883.04
SH30883.07
SH30883.01
76894-140EA
1724
USD
InStock
CA76894-140
CA76894-162
CA76894-152
76894-140
CA76894-160
76894-162
76894-152
76894-160
CA76894-106
76894-106
CA76894-124
76894-124
Sodium hydroxide solution 0.1 N (0.1 M), in single-use containers, HyClone™ for bioprocessing
Sodium hydroxide
HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean your chromatography resins.
- Manufactured in ISO 9001 and ISO 13485 certified facility.
- Lot-specific certificate of analysis (CoA) for each lot produced.
- Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
- 0.1 µm sterile filtered.
- All unit sizes are filled in single-use, ready-to-use bags.
- Raw materials are USP/NF and EP compendial grade.
HyClone™ NaOH in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 0.1 M (0.1 N), 0.5 M (0.5 N), and 1.0 M (1.0 N) sodium hydroxide concentrations up to 200 L.