Would you like to log in again?
BD MAX™ LBC Sample Buffer Tubes
Supplier: BD Diagnostics
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BD MAX™
443923
76906-232PK 157.88 USD InStock
76906-232
BD MAX™ LBC Sample Buffer Tubes
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
BD MAX™ liquid based cytology (LBC) sample buffer tubes.
  • Liquid based cytology specimens in ThinPrep® PreservCyt® solution
  • No re-capping step by lab technician after vortex
  • Post-testing, a sample buffer tube with a pierced cap may also be stored upright for up to 4 days at 2 to 30 °C
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty