Would you like to log in again?
BD MAX™ Urine Transport Kit
Supplier: BD Diagnostics
BD MAX™
443924
76906-234PK 346.58 USD InStock
76906-234
BD MAX™ Urine Transport Kit
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
BD MAX™ urine transport kit used for collection of male and female urine.
  • Male and female urine collection device, introduction of new pierceable caps
  • Eliminates re-capping step by lab technician and helps to reduce waste (1 cap/sample)
  • Post-testing, a sample buffer tube with a pierced cap may also be stored upright for up to 4 days at 2 to 30 °C
  • Transport stability at 2 to 30 ⁰C vs. UVE specimen collection kit (21
  • days vs. 5 days)
  • Elimination of Pre-warm step for urine sample
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty