Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DuPont™ Tyvek® Autoclavable Bowl Cover, with Steam Indicator, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products
DuPont™
BGT2A0103E BWLT2A30-2PLY6
76520-568CS 1467.4 USD InStock
76520-568 76918-244
DuPont™ Tyvek® Autoclavable Bowl Cover, with Steam Indicator, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.

  • 6" 2-ply edges resist punctures
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator
  • Autoclavable at 121 to 125 °C
  • Hydrophobic
  • Lint-free
  • Manufactured and packaged in a controlled cleanroom environment

Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilization. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.

After sterilization and after the outer Tyvek® bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed. Remaining in place over the bowl during setup, the cover minimizes microbial or particulate contamination risks.

Certifications: Certificates of Conformance sent with each shipment.

Packaging: Individually poly bagged.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty