Would you like to log in again?
VWR® Premium Laboratory Refrigerators
Supplier: VWR
VWR®
VWR-HC-SPL-23 VWR-HC-SPL-49 VWR-HC-PL-23 VWR-HC-PL-49
76919-388EA 11855.3 USD InStock
76919-388 76919-378 76919-386 76919-380
VWR® Premium Laboratory Refrigerators
Refrigerators
VWR® Premium Lab Refrigerators deliver next-level cooling to laboratory environments.
  • Energy saving VSC compressors
  • Microprocessor temperature controllers
  • Ultra-quiet operation
  • Fully array of alarms
  • Controller battery backup
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
Engineered with variable speed compressors (VSCs), these units feature ultra-quiet operation and significant energy savings. VSCs optimize energy consumption by self-adjusting to cooling demands. These compressors also deliver enhanced system performance and provide a longer lifespan than other compressor variations.
Enjoy the uniformity and speed of microprocessor temperature control and a full array of alarms to safeguard your products. Unit controllers also come with a battery backup to keep your data safe. Upgrade your laboratory environment with these premium refrigerators and welcome energy savings, noise reduction, smooth operation, and improved system performance to your work environment.
Certifications: ETL listed
*Select models ENERGY STAR certified: 76919-378, 76919-380, 76919-386 and 76919-388
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty