Would you like to log in again?
SteraMist SteraPak
Supplier: Tomi Environmental Solutions
S-PAK
76919-428EA 8772 USD InStock
76919-428
SteraMist SteraPak
Disinfection Systems
The all-in-one SteraPak places the power of SteraMist onto your back, delivering innovative disinfection that utilizes a rechargeable battery, cordless operation, and a new compact BIT solution bottle. 
  • The fully portable back-mounted SteraMist disinfection unit that offers unprecedented flexibility
  • Easily switch between battery and direct power
  • Delivers ionized Hydrogen Peroxide (iHP) in the most lightweight SteraMist system
  • For use with BIT-100 Solution
Perfect for maneuvering through small areas, such as attics and crawlspaces.
Delivery information: Includes SteraMist handheld applicator with 4’ cord, 1× SteraPak battery and charger, 32 oz DI water cartridge.
Order Now
ORDER
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty