Would you like to log in again?
SteraMist Total Disinfection Cart
Supplier: Tomi Environmental Solutions
CART-100
76919-434EA 39780 USD InStock
76919-434
SteraMist Total Disinfection Cart
Disinfection Systems
The SteraMist Disinfection Cart utilizes the SteraMist Surface Unit in a convenient hospital-grade cart.
Delivery information: Includes SteraMist® Surface Unit (SUR-103-110), durable shelved stainless steel transport cart, 2 gal. of BIT solution, Dräger® X-am 5100 Portable H2O2 Monitor, X-POWER 3580 air scrubber with dual carbon filters, MaxAir® system helmet with positive pressure air flow system, caution sign.
Order Now
ORDER
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty