Would you like to log in again?
SteraMist Surface Unit
Supplier: Tomi Environmental Solutions
SUR-103
76919-438EA 22440 USD InStock
76919-438
SteraMist Surface Unit
Disinfection Systems
The SteraMist Surface Unit is a portable, handheld point-and-spray disinfection and decontamination system.
  • Mobile for rapid deployment throughout a variety of spaces
  • Economical, non-corrosive, and easy to apply
  • Easily incorporated into daily cleaning protocols
  • Portable and rugged design for easy storage
  • 30’ removable applicator cord helps you reach a wide radius
  • Routine disinfection for large areas and surfaces
The SteraMist Surface Unit is the perfect solution for facility surface disinfection, bringing you handheld control with more movement and mobility around the treated area. 
Delivery information: Includes SteraMist handheld applicator, tool kit and 1 gal. DI water cartridge
Order Now
ORDER
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty