Would you like to log in again?
SteraMist Environment System
Supplier: Tomi Environmental Solutions
ENV-303-110
76919-446EA 88740 USD InStock
76919-446
SteraMist Environment System
Disinfection Systems
The SteraMist Environment System, the most intelligent mobile unit, delivers disinfection and decontamination with unparalleled scalability.
  • 3 removable 30’ cords and applicator stands
  • Intuitive HMI and custom cycle creation to meet all GMP requirements
  • Features remote operation and hand-held modes
  • Contains applicators able to treat areas of any size
Ideal for outbreaks and severe decontamination and deodorizing needs.
Delivery information: Includes 3× remote applicators (single point connectors with 30’ cords), 3× remote applicator stands and mounts, tool kit, 1 gal. DI water cartridge.
Order Now
ORDER
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty