Would you like to log in again?
VWR® Cleanroom Safety Shoes, Slip-On
Supplier: Avantor
VWR®
70D1059-48 70D1059-36 70D1059-37 70D1059-35 70D1059-39 70D1059-38 70D1059-45 70D1059-40 70D1059-44 70D1059-34 70D1059-43 70D1059-41 70D1059-42 70D1059-46
76944-206CS 650 USD InStock
76944-206 76944-208 76944-212 76944-202 76944-204 76944-190 76944-192 76944-186 76944-198 76944-210 76944-188 76944-200 76944-194 76944-196
VWR® Cleanroom Safety Shoes, Slip-On
Shoes and Boots
Premium comfort cleanroom and safety shoes.
  • Comfortable, form fitting, and lightweight design
  • Microfiber upper, with polyester fiber sewing
  • Mesh interior for breathability
  • Anti-static sole material
  • Polycarbonate toe cap
Premium cleanroom shoes designed with foot comfort in mind. These shoes feature a microfiber PU leather upper that is 1.8 mm thick. The mesh interior provides added breathability. Polyester fiber seams.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty