Would you like to log in again?
VWR® Max Protection High ESD Cleanroom Coveralls
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Max Protection
414004-314H 414004-685H 414004-310H 414004-311H 414004-312H 414004-313H 76311-056H 414004-320H 414004-5B6B-M 414004-319H 414004-686H 414004-306H 414004-321H 414004-308H 414004-684H 414004-315H 414004-5B6B-L 414004-316H 414004-317H 414004-5B6B-2XL 414004-5B6B-4XL 414004-5B6B-XL 414004-318H 414004-309H 414004-5B6B-3XL 414004-683H 414004-305H
76944-246CS 128.22 USD InStock
76944-246 76944-268 76944-248 76944-290 76944-252 76944-274 76944-254 76944-276 76944-292 76944-270 76944-294 76944-250 76944-272 76944-256 76944-278 76944-258 76944-280 76944-264 76944-242 76944-286 76944-266 76944-244 76944-288 76944-260 76944-282 76944-262 76944-284
VWR® Max Protection High ESD Cleanroom Coveralls
Disposable Garments
Cleanroom ESD coveralls are available in a variety of protective formats, including garments with attached hood, non-skid boots, or both.
  • Cleanroom coverall designed to provie high ESD properties
  • Lightweight, comfortable fit
VWR® Max Protection coveralls are made from 63 GSM SF and provide high ESD protection. All coveralls feature a front zipper closure, snap release on the leg, and serged seams. Elastic wrists and ankles are provided on garment without attached hood or boots.
Packaging: Coveralls are packaged in a case of 25.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty