Would you like to log in again?
PIP® Mission™ Indirect Vent Goggles
Supplier: Protective Industrial Products
PIP®
251-63-0520-RHB
76944-638CS 878.41 USD InStock
76944-638
PIP® Mission™ Indirect Vent Goggles
Goggles
Indirect vent goggles feature a green body, clear lens with FogLess® 3Sixty™ coating, and a Neoprene strap.
  • Indirect vent for liquid splash protection
  • Clear polycarbonate lens with anti-scratch and FogLess® 3Sixty™ coating
  • Adjustable neoprene strap provides a comfortable fit
  • Strap buckle for easy donning when wearing head protection
  • Fits over most prescription glasses
  • Comfortable TPR seal for comfort and extended use
Lightweight, semi-rimless design provides all-day user comfort. The sleek, sporty style with wrap-around lens protects the wearer against projectiles, impact, and liquid splash. FogLess® 3Sixty™ provides the ultimate anti-fog protection.
Certifications: ANSI Z87.1-2015 D3 Splash/Droplet compliant.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty