Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
G-Tek® VR-X™ Nylon Gloves with Polyurethane Coating
Supplier: Protective Industrial Products
G-Tek® VR-X™
33-VRX180/M 33-VRX180/XXL 33-VRX180/XS 33-VRX180/S 33-VRX180/L 33-VRX180/XL
76944-800DZ 48.19 USD InStock
76944-800 76944-802 76944-798 76944-808 76944-804 76944-806
G-Tek® VR-X™ Nylon Gloves with Polyurethane Coating
Gloves
Seamless knit nylon glove with polyurethane advanced barrier protection coating on full hand.
  • ESD safe gloves
  • Seamless construction offers increased comfort and breathability
  • Nylon blended shell, with an ultra-thin 18 gauge liner for exceptional dexterity
  • Liquid-proof, air leak tested
  • 4.0 AQL, similar to the AQL levels for industrial grade disposable gloves
  • Touchscreen compatible; operate a touch screen phone or device without removing gloves
  • Silicone-free
  • Knit wrist prevents dirt and debris from entering the glove
  • Color-coded hems for easy size identification
VR-X™ polyurethane coating provides advanced barrier protection. The treated coating takes on the surface properties of the glove liner, providing a tactile grip in wet, dry and slightly oily conditions. Ideal for applications that require dexterity for precise handling.
Ideal for handling and assembly of small to medium sharp parts and materials, manufacturing, inspection, shipping and packaging, and mechanical equipment maintenance and repair. Washable, resistant to water and ultraviolet light. Recommended wash cycle: maximum six cycles.
Certifications: Gloves provide antibacterial and antiviral barrier protection per ISO 20743:2013 test method and EN374-5. ESD safe, surface resistance per ESD S11.11 static dissipative range at 10⁸ ohms, 12% RH, 48hrs. @100volts.
Packaging: Gloves are packaged 12 dozen per case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty