Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MaxiFlex® Elite™ Anti-Static Nylon Gloves with Nitrile Coating
Supplier: Protective Industrial Products
MaxiFlex® Elite™
34-774B/XS 34-774B/L 34-774B/S 34-774B/XL 34-774B/XXL 34-774B/M
76944-810DZ 87.08 USD InStock
76944-810 76944-812 76944-820 76944-818 76944-814 76944-816
MaxiFlex® Elite™ Anti-Static Nylon Gloves with Nitrile Coating
Gloves
Anti-static seamless knit nylon glove with nitrile coated MicroFoam grip on palm and fingers.
  • Ultra light-weight nylon liner creates smooth and rounded fingertips maximizing sensitivity, dexterity and tactility
  • 30% thinner than traditional foam nitrile gloves
  • Touchscreen compatible; operate a touch screen phone or device without removing gloves
  • Patented nitrile MicroFoam coating offers 360° breathability
  • Minimal penetration (strike through), leaving only the soft liner resting against the skin
  • Silicone-free and latex-free
Weighing in at a significant 36% less and 30% thinner than traditional foam nitrile gloves MaxiFlex® Elite™ delivers a user experience that sets new standards. Comfort is further enhanced through their increased breathability, which is up to 40% more than similar gloves.
Certifications: Complies with EN 16350-2014 for electrostatic protective gloves. Also tested to EN 61340-2-3:2017 for achieving anti-static properties with vertical resistance.
Packaging: Gloves are packaged 12 dozen per case.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty