Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PIP® AR/FR Long Sleeve T-Shirts

Supplier: Protective Industrial Products
PIP®
385-FRLS-(GR)-3X 385-FRLS-(OR)-S 385-FRLS-(GR)-S 385-FRLS-(GR)-4X 385-FRLS-(LG)-XL 385-FRLS-(GR)-2X 385-FRLS-(NV)-3X 385-FRLS-(LG)-L 385-FRLS-(GR)-M 385-FRLS-(NV)-XL 385-FRLS-(OR)-2X 385-FRLS-(OR)-L 385-FRLS-(LG)-3X 385-FRLS-(LG)-M 385-FRLS-(OR)-3X 385-FRLS-(OR)-XL 385-FRLS-(GR)-XL 385-FRLS-(NV)-M 385-FRLS-(OR)-M 385-FRLS-(NV)-L 385-FRLS-(LG)-2X 385-FRLS-(GR)-L 385-FRLS-(NV)-2X 385-FRLS-(OR)-4X
76944-854CS 1695.63 USD InStock
76944-854 76944-876 76944-898 76944-866 76944-888 76944-878 76944-856 76944-868 76944-894 76944-872 76944-862 76944-884 76944-852 76944-874 76944-896 76944-886 76944-864 76944-858 76944-890 76944-880 76944-870 76944-892 76944-860 76944-882
PIP® AR/FR Long Sleeve T-Shirts
Workwear
ARC rated flame-resistant long sleeve T-shirts are constructed from 100% cotton.

  • 6.5 oz. 100% cotton interlock knit
  • Arc rating: ATPV 11.8 Cal/cm²
  • PPE Category: 2

AR/FR Long Sleeve T-shirts feature one chest pocket.

Certifications: Fabric meets requirements in accordance with ANSI/NFPA 2112-2012. Meets NFPA 70E, ASTM F1506.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty