Would you like to log in again?
Salam Adult Body Bag, Vinyl, Envelope Zipper, White
Supplier: Mopec
Salam
BE411-204-EZ
76945-182CS 397.08 USD InStock
76945-182
Salam Adult Body Bag, Vinyl, Envelope Zipper, White
Bags Storage Bags
Mopec in partnership with Salam International provide excellent body bags.
  • Ends are high frequency welded
  • Heavy-duty #7 zipper
  • 2 large, lockable pulls to secure pouch
  • Envelope zipper
  • 2 way zipper so pouch can open and close at any point
  • No sewing or welding on sides to increase strength
  • 100% leakproof
Materials: 7 to 9 mil vinyl
Dimensions: 94×36" (L×W)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty