Would you like to log in again?
APM Adult Body Bag, 4 - 8 Mil PVC, Center/Envelope Zipper, White or Yellow
Supplier: Mopec
APM
BE802E-Y-XL BE802E-LW BE802-XL BE802-LW BE802E-XL BE802E BE802 BE802E-Y
76945-302CS 969.22 USD InStock
76945-302 76945-310 76945-300 76945-306 76945-304 76945-308 76945-298 76945-296
APM Adult Body Bag, 4 - 8 Mil PVC, Center/Envelope Zipper, White or Yellow
Bags Storage Bags
Mopec in partnership with American Professional Manufacturing provide excellent body bags.
  • Custom-made, lockable slider for easy opening and closing
  • Dielectrically sealed seams
  • Available with center or envelope zipper
  • Color: white or yellow
  • Leak resistance
Materials: 4 to 8 mil PVC
Dimensions: 94×36" or 94×48" (L×W)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty