Would you like to log in again?
APM Young Body Bag, 8 Mil PVC, Center Zipper, White
Supplier: Mopec
APM
BE804 BE807 BE805
76945-316CS 263.92 USD InStock
76945-316 76945-318 76945-320
APM Young Body Bag, 8 Mil PVC, Center Zipper, White
Bags Storage Bags
Mopec in partnership with American Professional Manufacturing provide excellent body bags.
  • Custom-made, lockable slider for easy opening and closing
  • Dielectrically sealed seams
  • Center zipper
  • Color: white
  • Leak resistance
Materials: 8 mil PVC
Dimensions: Various sizes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty