Would you like to log in again?
Control Slides, Special Stains, Iron (Ferric) In Bone Marrow, Spleen, Liver
Supplier: Mopec
SC818-20 SC818-100 SC818-10
76947-926CS 286.79 USD InStock
76947-926 76947-924 76947-922
Control Slides, Special Stains, Iron (Ferric) In Bone Marrow, Spleen, Liver
Slides Microscopy Control Slides
All positive control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Packaging: Case of 10 contains 10 unstained slides and 1 stained slide. Case of 20 contains 20 unstained slides and 2 stained slides. Case of 100 contains 98 unstained slides and 2 stained slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty