Would you like to log in again?
Control Slides, Tissue Microarray, Normal Human Tissue, 16 Tissue Types
Supplier: Mopec
SC984-25
76948-262CS 3056.18 USD InStock
76948-262
Control Slides, Tissue Microarray, Normal Human Tissue, 16 Tissue Types
Slides Microscopy Control Slides
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 μm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Tissue type: chosen from Liver, Lung, Kidney, Colon, Tonsil, Skin, Prostate, Breast, Lymph Node, Uterus, Testis, Placenta, Thyroid, Stomach, and Pancreas.
Ordering information: Case of 25 contains 24 unstained slides and 1 stained slide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty