Would you like to log in again?
Coverslips
Supplier: Mopec
SL024-50-10 SL024-60-10 SL024-55-20 SL024-55-10 SL024-50-20
76948-282CS 215.76 USD InStock
76948-282 76948-284 76948-280 76948-286 76948-288
Coverslips
Cover Glasses
Mopec coverslips are made with borosilicate glass.
  • Borosilicate glass, #1 thickness, 12 mm in diameter
  • Will not stick or auto-fluoresce
  • Clean, uniform surface
  • Ideal for automated coverslipping
Available in 1 or 2 ounce volumes.
Packaging: The coverslips are packaged 10 boxes per case, with 100 or 200 pieces per box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty