Would you like to log in again?
Microscope Slides, Regular Non-Charged, 90° Corners
Supplier: Mopec
SL090-TN-CS SL090-YL-CS SL090-YL-BX SL090-PK-BX SL090-BL-BX SL090-GR-BX SL090-TN-BX SL090-GR-CS SL090-PK-CS SL090-WH-CS SL090-BL-CS SL090-LI-CS SL090-LI-BX SL090-WH-BX SL090-PE-BX SL090-PE-CS
76948-350BX 25.14 USD InStock
76948-350 76948-326 76948-348 76948-338 76948-328 76948-344 76948-322 76948-334 76948-324 76948-346 76948-336 76948-340 76948-352 76948-330 76948-342 76948-332
Microscope Slides, Regular Non-Charged, 90° Corners
Slides Microscope Slides
High-quality, uncharged microscope slides with color coding.
  • 1×3" slides, 1 mm thickness
  • 100% guaranteed against sticking and contamination
  • 90 degree corners
  • Uniform flat paint for perfect barcodes
Slides are available in white, blue, pink, yellow, green, tan, peach, and lilac. The packaging options include 1 box of 72 slides and 1 case of 1440 slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty