Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microscope Slides, Charged, Inkjet, 45° Corners

Supplier: Mopec
SL245-WH-CS SL245-PE-CS SL245-LI-CS SL245-BL-CS SL245-YL-BX SL245-GR-CS SL245-PK-CS SL245-LI-BX SL245-TN-BX SL245-PK-BX SL245-YL-CS SL245-TN-CS SL245-WH-BX SL245-GR-BX SL245-PE-BX SL245-BL-BX
76948-418BX 44.33 USD InStock
76948-418 76948-436 76948-426 76948-448 76948-438 76948-428 76948-432 76948-444 76948-422 76948-434 76948-446 76948-424 76948-440 76948-430 76948-420 76948-442
Microscope Slides, Charged, Inkjet, 45° Corners
Slides Microscope Slides
High-quality, charged microscope slides with color coding.

  • For Inkjet
  • 1×3" slides, 1mm thickness
  • 100% guaranteed against sticking and contamination
  • 45 degree corners
  • Uniform flat paint for perfect barcodes

Slides are available in white, blue, pink, yellow, green, tan, peach, and lilac.

Packaging: The packaging options include 1 box of 72 slides and 1 case of 1440 slides.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty