Would you like to log in again?
Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 22 mm, Available in Multiple Colors
Supplier: Mopec
SP101-OR SP101-PK SP101-RD SP101-WH SP101-BL SP101-GN SP101-YE SP101-PR
76948-506EA 71.6 USD InStock
76948-506 76948-508 76948-502 76948-504 76948-498 76948-510 76948-500 76948-496
Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 22 mm, Available in Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for leica (except CM1950 model), TBS, and tanner cryostats
  • 22 mm
  • Some variations also available in waffle pattern
Available in red, orange, yellow, green, blue, purple, pink and white.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty