Would you like to log in again?
Circular Specimen Chucks For Leica, TBS, And Tanner Cryostats, 30 mm, Available In Multiple Colors
Supplier: Mopec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SP103-GN SP103-PR SP103-WH SP103W-GN SP103W-PK SP103W-BL SP103-PK SP103-BL SP103-YE SP103W-WH SP103W-RD SP103-RD SP103W-PR SP103W-OR SP103W-YE SP103-OR
76948-590EA 140.36 USD InStock
76948-590 76948-580 76948-592 76948-528 76948-536 76948-538 76948-586 76948-542 76948-532 76948-588 76948-534 76948-582 76948-594 76948-540 76948-584 76948-530
Circular Specimen Chucks For Leica, TBS, And Tanner Cryostats, 30 mm, Available In Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner Cryostats
  • 30 mm
  • Some variations also available in waffle pattern
Available in red, orange, yellow, green, blue, purple, pink and white
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty