Would you like to log in again?
Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 40 mm, Available in Multiple Colors
Supplier: Mopec
SP104W-GN SP104W-PK SP104W-WH SP104-WH SP104W-RD SP104-GN SP104-BL SP104-OR SP104-PR SP104-RD SP104W-OR SP104W-YE SP104-PK SP104W-PR SP104-YE SP104W-BL
76948-606EA 170.17 USD InStock
76948-606 76948-608 76948-546 76948-602 76948-558 76948-548 76948-604 76948-598 76948-554 76948-544 76948-610 76948-556 76948-600 76948-550 76948-552 76948-596
Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 40 mm, Available in Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for leica (except CM1950 model), TBS, and tanner cryostats
  • 40 mm
  • Some variations also available in waffle pattern
Available in red, orange, yellow, green, blue, purple, pink and white.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty