Would you like to log in again?
Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 55 mm, Available in Multiple Colors
Supplier: Mopec
SP105-PK SP105-BL SP105-GN SP105-OR SP105-RD SP105-PR SP105-WH SP105-YE
76948-570EA 135.78 USD InStock
76948-570 76948-568 76948-564 76948-566 76948-560 76948-572 76948-562 76948-574
Circular Specimen Chucks for Leica, TBS, and Tanner Cryostats, 55 mm, Available in Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for Leica (except CM1950 model), TBS, and Tanner cryostats
  • Some variations also available in waffle pattern
Available in red, orange, yellow, green, blue, purple, pink and white.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty