Would you like to log in again?
Specimen Chucks for QS And Microm Cryostats, 40 mm, Available in Multiple Colors
Supplier: Mopec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SP134W-PK SP134-RD SP134W-PR SP134-WH SP134W-BL SP134W-GN SP134-YE SP134W-YE SP134W-RD SP134-GN SP134-OR SP134-PR SP134-BL SP134W-OR SP134W-WH SP134-PK
76948-790EA 170.17 USD InStock
76948-790 76948-766 76948-788 76948-756 76948-800 76948-768 76948-802 76948-758 76948-762 76948-796 76948-764 76948-798 76948-770 76948-792 76948-760 76948-794
Specimen Chucks for QS And Microm Cryostats, 40 mm, Available in Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for QS and Microm Cryostats
Available in red, orange, yellow, green, blue, purple, pink and white.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty