Would you like to log in again?
Circular Specimen Chucks For SHANDON Cryostats, 30 mm, Available In Multiple Colors
Supplier: Mopec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SP143-PR SP143W-PR SP143W-WH SP143W-PK SP143-RD SP143W-BL SP143-GN SP143-WH SP143-OR SP143-PK SP143-YE SP143W-YE SP143W-OR SP143W-RD SP143W-GN SP143-BL
76948-818EA 140.36 USD InStock
76948-818 76948-808 76948-814 76948-836 76948-848 76948-804 76948-816 76948-838 76948-806 76948-810 76948-844 76948-812 76948-846 76948-850 76948-840 76948-842
Circular Specimen Chucks For SHANDON Cryostats, 30 mm, Available In Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for SHANDON Cryostats
  • 30 mm
Available in red, orange, yellow, green, blue, purple, pink and white
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty