Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Circular Specimen Chucks for SHANDON Cryostats, 30 mm, Available in Multiple Colors

Supplier: Mopec

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

SP144-YE SP144-OR SP144-PR SP144-BL SP144W-OR SP144W-BL SP144W-PR SP144W-GN SP144W-RD SP144-WH SP144W-WH SP144W-PK SP144-GN SP144-PK SP144W-YE SP144-RD
76948-858EA 234.34 USD InStock
76948-858 76948-826 76948-828 76948-854 76948-832 76948-822 76948-866 76948-834 76948-856 76948-824 76948-862 76948-852 76948-830 76948-820 76948-864 76948-860
Circular Specimen Chucks for SHANDON Cryostats, 30 mm, Available in Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.

  • Round, aluminum specimen chucks
  • Suitable for SHANDON cryostats
  • 40 mm

Available in red, orange, yellow, green, blue, purple, pink and white.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty