Would you like to log in again?
Microscope Slide Mailers, Holds 5 Microscope Slides, Translucent, 100/Box
Supplier: Mopec
SP235
76948-902BX 46.39 USD InStock
76948-902
Microscope Slide Mailers, Holds 5 Microscope Slides, Translucent, 100/Box
Slide Mailers
Translucent mailer for microscope slides.
  • Snap top closure
  • 100/box
Slide mailers are designed to hold five (5) 1×3" standard microscope slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty