Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LISA Safety Waste Cap

Supplier: S.C.A.T.

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

LISA
350090 350045 350055 350063 350053 450060 350110 450045 350121 450120 350120 450110 350095 350050 350100 350060 350065 350051 450100 350070 350083 350099
76948-928EA 269.56 USD InStock
76948-928 76948-946 76948-958 76948-936 76948-926 76948-948 76948-938 76948-942 76948-932 76948-976 76948-954 76948-944 76948-934 76948-956 76948-978 76948-982 76948-950 76948-940 76948-974 76948-952 76948-930 76948-980
LISA Safety Waste Cap
Closures Safety Caps and Safety Waste Caps
The first modular system for HPLC waste. Workplace safety and environmental protection with the proven technology, providing maximum flexibility. Simply add connections and components individually as required. The modular system grows with the user, allowing solvents, acids and alkalis to be collected safely.

  • Satellite blind plug extends and attaches various components, such as a funnel
  • Tube connector allows connection of larger tubes with 5 to 11.5 mm Int.Ø
  • Satellite made of PTFE for optimal chemical resistance
  • 360° freely rotatable screw cap; easy changing of the container without twisting the tubing
  • Fittings feature an improved design for easier connection of capillaries with 1.6, 2.3, or 3.2 mm Ext.Ø
  • It exhaust filters block harmful vapours with activated carbon, ensuring safe pressure equalisation in the waste containers

The huge scope of delivery of fittings and blind plugs as well as accessories such as exhaust filters and funnels ensures that the docking system is ready for immediate use. The basic module includes a safety waste cap LISA with selected thread size. Included in the scope of delivery are a further 23 accessories. In addition, a variety of other accessories such as a funnel or an exhaust air filter can be docked to the safety waste cap LISA V3.0.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty