Would you like to log in again?
Quick-Lock Connectors
Supplier: S.C.A.T.
360189 360183
76948-964PK 117.18 USD InStock
76948-964 76948-966
Quick-Lock Connectors
Fittings and Connectors
Easy and safe connection of capillary tubing and SCAT safety caps using quick couplings.
  • Made of chemically resistant glass-filled polypropylene, PEEK® materials
  • Leak protection thanks to integrated shut-off valves
  • Closed system for drip-free disconnection with very low air entrapment
  • Minimizes contamination and no need to purge air bubbles from the system
  • Suitable for capillaries with outer diameter 1.6, 2.3, and 3.2 mm
  • User-friendly, ergonomic design, enables safe connection
  • Up to vacuum 15 psi, 1,0 bar pressure resistance
Connect and disconnect with a 'click', without dripping or air bubbles. The valve closes automatically when disconnected. Thanks to the included PFA fittings, all common hose diameters can be connected.
The quick-lock connector sets are suitable for connecting 2 capillaries (set 1) or one capillary and one SCAT safety cap (set 2).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty