Would you like to log in again?
Plain Microscope Slides
Supplier: Four E's USA
K2106160 K2106161
76970-066BX 9.33 USD InStock
76970-066 76970-064
Plain Microscope Slides
Slides Microscope Slides
Four E's microscope slides made from high quality soda glass or super white glass.
  • Pre-cleaned, ready to use
  • Grounded edges, 45° corners
  • Autoclavable
Suitable for traditional identification labeling, these plain slides are manufactured using an advanced grinding process to ensure uniform pieces and consistency in flatness and smoothness. The polished edges and corners greatly reduce the risk of scratches and infection during use and handling of the slides.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty