Would you like to log in again?
Color Frosted Microscope Slides
Supplier: Four E's USA
K2106169 K2106170 K2106171 K2106168
76970-074BX 14.62 USD InStock
76970-074 76970-072 76970-078 76970-076
Color Frosted Microscope Slides
Slides Microscope Slides
Four E's color frosted microscope slides made from premium super white glass for ultra clarity with higher light transmittance.
  • Pre-cleaned, ready to use
  • Color frosted on one end, one side
  • Grounded edges, 45° corners
  • Autoclavable
  • Colors available: White, green, blue and yellow
The colorfast opaque coating is resistant to commonly used laboratory solvents. The chromatic integrity stays intact even through routine H&E staining.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty