Would you like to log in again?
Body Bag, GOV/DOD Design Disaster Bag, Envelope Zipper
Supplier: Mopec
BE167-EZ
76970-100EA 179.6 USD InStock
76970-100
Body Bag, GOV/DOD Design Disaster Bag, Envelope Zipper
Bags Storage Bags
Designed to be similar to Dept of Defense (DOD) Specification NSN: 9930-01-331-6244 in strength and durability, the Mopec BE167-EZ contains the following deviations in order to improve production time during the height of the COVID-19 crisis.
  • Envelope zipper
  • 100% leakproof
  • Black
  • Leak proof heat sealed seams
  • 6 handles
Material: Heavy duty, 18 mil PVC supported vinyl.
Dimensions: 36×96"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty